AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride) ≥98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride, AEBSF hydrochloride

BIMB2003-1G BIMB2003-500MG BIMB2003-5G
APOSBIMB2003-1GEA 132 EUR
APOSBIMB2003-1G APOSBIMB2003-500MG APOSBIMB2003-5G
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride) ≥98%
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)

An irreversible serine protease inhibitor with highwater solubility.


  • Molecular biology grade

Formule: C₈H₁₁ClFNO₂S
MW: 239.69 g/mol
Smeltpunt: 182…184 °C (ontbinding)
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00132962
CAS nummer: 30827-99-7
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR