2'-Fluoro-2'-deoxyinosine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

BIMN2011-100MG BIMN2011-1G BIMN2011-250MG
APOSBIMN2011-100MGEA 103 EUR
APOSBIMN2011-100MG APOSBIMN2011-1G APOSBIMN2011-250MG
2'-Fluoro-2'-deoxyinosine
2'-Fluoro-2'-deoxyinosine
Formule: C₁₀H₁₁FN₄O₄
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD09750860
CAS nummer: 80049-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR