Amino-PEG6-t-butyl ester

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

BIPG1075-250MG BIPG1075-100MG BIPG1075-500MG
APOSBIPG1075-250MGEA 125 EUR
APOSBIPG1075-250MG APOSBIPG1075-100MG APOSBIPG1075-500MG
Amino-PEG6-t-butyl ester
Amino-PEG6-t-butyl ester
Formule: C₁₉H₃₉NO₈
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD13184944
CAS nummer: 1286281-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR