Bromo-PEG3-t-butyl ester

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

BIPG1349-250MG BIPG1349-100MG
APOSBIPG1349-250MGEA 67.2 EUR
APOSBIPG1349-250MG APOSBIPG1349-100MG
Bromo-PEG3-t-butyl ester
Bromo-PEG3-t-butyl ester
Formule: C₁₃H₂₅BrO₅
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD22574821
CAS nummer: 782475-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR