Fmoc-N-amido-PEG24-acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

BIPG1407-100MG
APOSBIPG1407-100MGEA 402 EUR
APOSBIPG1407-100MG
Fmoc-N-amido-PEG24-acid
Fmoc-N-amido-PEG24-acid
Formule: C₆₆H₁₁₃NO₂₈
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD11041159
CAS nummer: 2170484-59-8

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR