Tiamulin fumarate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

BIT0157-1G BIT0157-250MG BIT0157-5G
APOSBIT0157-1GEA 80.4 EUR
APOSBIT0157-1G APOSBIT0157-250MG APOSBIT0157-5G
Tiamulin fumarate
Tiamulin fumarate

  • Antibodies

Formule: C₃₂H₅₁NO₈S
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00145407
CAS nummer: 55297-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR