Calcium bromide, anhydrous 99,5%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

IN1378-100G IN1378-250G
APOSIN1378-100GEA 110 EUR
APOSIN1378-100G APOSIN1378-250G
Calcium bromide, anhydrous 99,5%
Calcium bromide
Formule: Br₂Ca
MW: 199.89 g/mol
Kookpunt: 806…812 °C
Smeltpunt: 730 °C (ontbinding)
Dichtheid: 3.353 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010902
CAS nummer: 7789-41-5
EINECS: 232-164-6
Merck Index: 12,01693

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR