Copper 99.999%, shot 4 - 6 mm

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

IN1531-250G
APOSIN1531-250GEA 1390 EUR
APOSIN1531-250G
Copper 99.999%, shot 4 - 6 mm
Copper
Formule: Cu
Kookpunt: 2595 °C
Smeltpunt: 1083 °C
Dichtheid: 8,94 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010965
CAS nummer: 7440-50-8
EINECS: 231-159-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR