Copper selenide 99.99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

IN1585-100G IN1585-25G
APOSIN1585-100GEA 622 EUR
APOSIN1585-100G APOSIN1585-25G
Copper selenide 99.99%
Copper selenide
Formule: CuSe
Dichtheid: 5,99 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00016058
CAS nummer: 1317-41-5
EINECS: 215-272-8
UN: 3283
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR