Copper selenide 99.999%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

IN1588-25G IN1588-100G
APOSIN1588-25GEA 319 EUR
APOSIN1588-25G APOSIN1588-100G
Copper selenide 99.999%
Copper selenide
Formule: CuSe
Dichtheid: 5,99 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00016058
CAS nummer: 1317-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR