Gallium 99.9999%, ingot

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

IN1855-10G IN1855-50G IN1855-100G
APOSIN1855-10GEA 256 EUR
APOSIN1855-10G APOSIN1855-50G APOSIN1855-100G
Gallium 99.9999%, ingot
Gallium
Formule: Ga
Kookpunt: 2403 °C/760mmHg
Dichtheid: 5,91 g/cm³ (25 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00134045
CAS nummer: 7440-55-3
UN: 2803
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR