Neodymium(III) acetate hydrate 99,9%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Neodymium(III) acetate xhydrate, Neodymium triacetate hydrate

IN2623-50G IN2623-250G
APOSIN2623-50GEA 110 EUR
APOSIN2623-50G APOSIN2623-250G
Neodymium(III) acetate hydrate 99,9%
Neodymium(III) acetate hydrate
Formule: C₆H₁₁NdO₇
MW: 339.39 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150120
CAS nummer: 334869-71-5
EINECS: 228-244-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR