Nickel(II) dibutyldithiocarbamate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

IN2708-25G
APOSIN2708-25GEA 23.4 EUR
APOSIN2708-25G
Nickel(II) dibutyldithiocarbamate
Nickel(II) dibutyldithiocarbamate
Formule: C₉H₁₉NS₂
Smeltpunt: 89 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00067273
CAS nummer: 13927-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR