Rhenium 99.999%, powder –22 mesh

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

IN2992-1G IN2992-5G
APOSIN2992-1GEA 145 EUR
APOSIN2992-1G APOSIN2992-5G
Rhenium 99.999%, powder –22 mesh
Rhenium
Formule: Re
Kookpunt: 5900 °C
Smeltpunt: 3180 °C
Dichtheid: 21,04 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011195
CAS nummer: 7440-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR