2-Aminopyridine 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR0015-1KG OR0015-100G OR0015-500G
APOSOR0015-1KGEA 109 EUR
APOSOR0015-1KG APOSOR0015-100G APOSOR0015-500G
2-Aminopyridine 99%
2-Aminopyridine
Formule: C₅H₆N₂
Kookpunt: 204…211 °C
Smeltpunt: 54…60 °C
Vlampunt: 92 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006312
CAS nummer: 504-29-0
EINECS: 207-988-4
UN: 2671
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR