Bis(4-chlorophenyl)disulphide 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR0016-10G OR0016-50G
APOSOR0016-10GEA 26.3 EUR
APOSOR0016-10G APOSOR0016-50G
Bis(4-chlorophenyl)disulphide 98%
Bis(4-chlorophenyl)disulphide
Formule: C₁₂H₈Cl₂S₂
Smeltpunt: 71…74 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013642
CAS nummer: 1142-19-4
EINECS: 214-531-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR