Ethyl-2-(hydroxyimino)-3-oxobutanoate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR015018-1G OR015018-25G OR015018-5G OR015018-250MG OR015018-500MG OR015018-10G
APOSOR015018-1GEA 81.8 EUR
APOSOR015018-1G APOSOR015018-25G APOSOR015018-5G APOSOR015018-250MG APOSOR015018-500MG APOSOR015018-10G
Ethyl-2-(hydroxyimino)-3-oxobutanoate
Ethyl-2-(hydroxyimino)-3-oxobutanoate
Formule: C₆H₉NO₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00039517
CAS nummer: 5408-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR