(S)-(+)-2-Aminobutyric acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: H-Abu-OH, L-alpha-Amino-N-Butyric acid, L-2-Aminobutyric acid, (S)-2-Aminobutanoic acid

OR01636-100G OR01636-25G OR01636-5G
APOSOR01636-100GEA 36.6 EUR
APOSOR01636-100G APOSOR01636-25G APOSOR01636-5G
(S)-(+)-2-Aminobutyric acid
(S)-(+)-2-Aminobutyric acid
Formule: C₄H₉NO₂
MW: 103.12 g/mol
Smeltpunt: 280 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064415
CAS nummer: 1492-24-6
EINECS: 216-083-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR