5-(2,4-Dichlorophenyl)-4H-pyrazole-3-carboxylic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR01715-500MG
APOSOR01715-500MGEA 162 EUR
APOSOR01715-500MG
5-(2,4-Dichlorophenyl)-4H-pyrazole-3-carboxylic acid
5-(2,4-Dichlorophenyl)-4H-pyrazole-3-carboxylic acid
Formule: C₁₀H₆Cl₂N₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06245447
CAS nummer: 904815-22-1

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR