Ethyl 2,6-dichlorobenzoate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR017267-1G OR017267-25G OR017267-5G
APOSOR017267-1GEA 35.1 EUR
APOSOR017267-1G APOSOR017267-25G APOSOR017267-5G
Ethyl 2,6-dichlorobenzoate
Ethyl-2,6-dichlorobenzoate
Formule: C₉H₈Cl₂O₂
MW: 219.07 g/mol
Kookpunt: 264…265 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01313702
CAS nummer: 81055-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR