4-Aminobutyric acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: GABA, γ-Aminobutyric acid

OR01972-100G OR01972-500G
APOSOR01972-100GEA 90.6 EUR
APOSOR01972-100G APOSOR01972-500G
4-Aminobutyric acid
4-Aminobutyric acid
Formule: C₄H₉NO₂
MW: 103.12 g/mol
Kookpunt: 248
Smeltpunt: 195 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008226
CAS nummer: 56-12-2
EINECS: 200-258-6
Merck Index: 13,00429

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR