2,4-Dimethoxyphenyl isothiocyanate 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR0408-5G OR0408-1G
APOSOR0408-5GEA 122 EUR
APOSOR0408-5G APOSOR0408-1G
2,4-Dimethoxyphenyl isothiocyanate 97%
2,4-Dimethoxyphenyl isothiocyanate
Formule: C₉H₉NO₂S
Kookpunt: 166…167 °C/6mmHg
Smeltpunt: 51…53 °C
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00041058
CAS nummer: 33904-03-9
UN: 2923
ADR: 8+6.1,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR