Sodium dicyanoamide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR0738-5G
APOSOR0738-5GEA 22 EUR
APOSOR0738-5G
Sodium dicyanoamide
Sodium dicyanoamide
Formule: C₂N₃Na
Smeltpunt: > 300 °C
MDL Nummer: MFCD00003535
CAS nummer: 1934-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR