Malonic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 1,3-Propanedioic acid

OR0786-100G OR0786-1KG OR0786-500G
APOSOR0786-100GEA 22 EUR
APOSOR0786-100G APOSOR0786-1KG APOSOR0786-500G
Malonic acid
Malonic acid
Formule: C₃H₄O₄
MW: 104.06 g/mol
Kookpunt: 387
Smeltpunt: 132…136 °C (ontbinding)
Dichtheid: 1.62 g/cm³ (25 °C)
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002707
CAS nummer: 141-82-2
EINECS: 205-503-0
Merck Index: 13,05732

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR