5-(4-Hydroxypiperidino)-2-thiophenecarbaldehyde

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR0900-5G
APOSOR0900-5GEA 231 EUR
APOSOR0900-5G
5-(4-Hydroxypiperidino)-2-thiophenecarbaldehyde
5-(4-Hydroxypiperidino)-2-thiophenecarbaldehyde
Formule: C₁₀H₁₃NO₂S
Smeltpunt: 137…139 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03425728
CAS nummer: 207290-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR