1-[2-(1H-Pyrrol-1-yl)ethyl]piperazine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR0959-250MG OR0959-1G OR0959-5G
APOSOR0959-250MGEA 32.2 EUR
APOSOR0959-250MG APOSOR0959-1G APOSOR0959-5G
1-[2-(1H-Pyrrol-1-yl)ethyl]piperazine
1-[2-(1H-Pyrrol-1-yl)ethyl]piperazine
Formule: C₁₀H₁₇N₃
Kookpunt: 96 °C (0,8 mm Hg)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03412127
CAS nummer: 688763-20-4

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR