1,3-Dichloorpropaan 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR0973-25G OR0973-100G OR0973-250G
APOSOR0973-25GEA 45.4 EUR
APOSOR0973-25G APOSOR0973-100G APOSOR0973-250G
1,3-Dichloorpropaan 99%
1,3-Dichloorpropaan
Formule: ClH₂CCH₂CH₂Cl
Kookpunt: 120 °C
Dichtheid: 1,19
Vlampunt: 21 °C
MDL Nummer: MFCD00000999
CAS nummer: 142-28-9
UN: 1933
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR