N-Methylanthranilic acid 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR10023-1G OR10023-5G
APOSOR10023-1GEA 22 EUR
APOSOR10023-1G APOSOR10023-5G
N-Methylanthranilic acid 99%
N-Methylanthranilic acid
Formule: C₈H₉NO₂
Smeltpunt: 170…172 °C
MDL Nummer: MFCD00002424
CAS nummer: 119-68-6

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR