3-(Boc-amino)phenylboronic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR10346-1G OR10346-5G OR10346-25G
APOSOR10346-1GEA 62.9 EUR
APOSOR10346-1G APOSOR10346-5G APOSOR10346-25G
3-(Boc-amino)phenylboronic acid 97%
3-(Boc-amino)phenylboronic acid
Formule: C₁₁H₁₆BNO₄
Smeltpunt: 168 °C
MDL Nummer: MFCD03411945
CAS nummer: 380430-68-2

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR