5-Bromo-1-methylimidazole

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 5-Bromo-1-methyl-1H-imidazole

OR10368-25G OR10368-5G OR10368-100G
APOSOR10368-25GEA 64.3 EUR
APOSOR10368-25G APOSOR10368-5G APOSOR10368-100G
5-Bromo-1-methylimidazole
5-Bromo-1-methylimidazole
Formule: C₄H₅BrN₂
MW: 161 g/mol
Kookpunt: 120 °C/15mmHg
Smeltpunt: 40…41 °C
Vlampunt: 82 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01632218
CAS nummer: 1003-21-0
UN: 3335
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR