4-(Methylsulphinylphenyl)boronic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR10559-1G OR10559-100MG OR10559-250MG OR10559-500MG
APOSOR10559-1GEA 356 EUR
APOSOR10559-1G APOSOR10559-100MG APOSOR10559-250MG APOSOR10559-500MG
4-(Methylsulphinylphenyl)boronic acid
4-(Methylsulphinylphenyl)boronic acid
Formule: C₇H₉BO₃S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02093071
CAS nummer: 166386-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR