Diethyl 4-carboxyphenylphosphonate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR10696-1G
APOSOR10696-1GEA 380 EUR
APOSOR10696-1G
Diethyl 4-carboxyphenylphosphonate
Diethyl 4-carboxyphenylphosphonate
Formule: C₁₁H₁₅O₅P
MW: 258.208 g/mol
Smeltpunt: 105…107 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01872486
CAS nummer: 1527-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR