Methylenediphosphonic acid 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Medronic acid

OR10789-1G OR10789-5G
APOSOR10789-1GEA 27.8 EUR
APOSOR10789-1G APOSOR10789-5G
Methylenediphosphonic acid 98%
Methylenediphosphonic acid
Formule: CH₆O₆P₂
MW: 176 g/mol
Smeltpunt: 197…204 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00002139
CAS nummer: 1984-15-2
EINECS: 217-851-0
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR