5-Chloroindole-2-carboxylic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR10933-25G
APOSOR10933-25GEA 99.3 EUR
APOSOR10933-25G
5-Chloroindole-2-carboxylic acid
5-Chloroindole-2-carboxylic acid
Formule: C₉H₆ClNO₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00005613
CAS nummer: 10517-21-2
EINECS: 234-050-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR