5-Bromo-N-methoxy-N-methyl-2-furamide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR110852-1G OR110852-500MG
APOSOR110852-1GEA 366 EUR
APOSOR110852-1G APOSOR110852-500MG
5-Bromo-N-methoxy-N-methyl-2-furamide
5-Bromo-N-methoxy-N-methyl-2-furamide
Formule: C₇H₈BrNO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD16104416
CAS nummer: 179055-22-2

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR