1,3-Bis(3,4-dichlorophenyl)urea

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR11101-1G OR11101-5G OR11101-500MG
APOSOR11101-1GEA 103 EUR
APOSOR11101-1G APOSOR11101-5G APOSOR11101-500MG
1,3-Bis(3,4-dichlorophenyl)urea
1,3-Bis(3,4-dichlorophenyl)urea
Formule: C₁₃H₈Cl₄N₂O
Smeltpunt: 281…282 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00171464
CAS nummer: 4300-43-0
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR