Ethyl 3-bromo-4-hydroxy-5-nitrobenzoate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR111201-1G OR111201-5G
APOSOR111201-1GEA 106 EUR
APOSOR111201-1G APOSOR111201-5G
Ethyl 3-bromo-4-hydroxy-5-nitrobenzoate
Ethyl 3-bromo-4-hydroxy-5-nitrobenzoate
Formule: C₉H₈BrNO₅
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD22383758
CAS nummer: 141778-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR