1-Bromo-2-methoxyethane 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR11137-100G OR11137-500G OR11137-1KG
APOSOR11137-100GEA 80.4 EUR
APOSOR11137-100G APOSOR11137-500G APOSOR11137-1KG
1-Bromo-2-methoxyethane 98%
1-Bromo-2-methoxyethane
Formule: BrH₂CCH₂OCH₃
Kookpunt: 109…110 °C
Dichtheid: 1,478 g/cm³ (25 °C)
Vlampunt: 28 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000236
CAS nummer: 6482-24-2
EINECS: 229-339-4
UN: 3271
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR