(4-Amino-3,5-dibromophenyl)acetic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR111426-1G
APOSOR111426-1GEA 42.4 EUR
APOSOR111426-1G
(4-Amino-3,5-dibromophenyl)acetic acid
(4-Amino-3,5-dibromophenyl)acetic acid
Formule: C₈H₇Br₂NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD20529161
CAS nummer: 191869-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR