5'-Chloro-2'-hydroxyacetophenone 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 1-(5-Chloro-2-hydroxyphenyl)ethan-1-one, 1-(5-Chloro-2-hydroxyphenyl)ethanone

OR1115-500G
APOSOR1115-500GEA 372 EUR
APOSOR1115-500G
5'-Chloro-2'-hydroxyacetophenone 98%
5'-Chloro-2'-hydroxyacetophenone
Formule: C₈H₇ClO₂
MW: 170.6
Kookpunt: 126…128 °C/28mmHg
Smeltpunt: 54…56 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00067788
CAS nummer: 1450-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR