3-Nitrobenzenesulphonyl chloride 96%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR11222-25G OR11222-5G OR11222-500G
APOSOR11222-25GEA 125 EUR
APOSOR11222-25G APOSOR11222-5G APOSOR11222-500G
3-Nitrobenzenesulphonyl chloride 96%
3-Nitrobenzenesulphonyl chloride
Formule: C₆H₄ClNO₄S
Smeltpunt: 61…62 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007435
CAS nummer: 121-51-7
EINECS: 204-476-2
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR