2-Amino-4,6-dichloropyrimidine 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR11290-100G OR11290-25G OR11290-500G
APOSOR11290-100GEA 24.9 EUR
APOSOR11290-100G APOSOR11290-25G APOSOR11290-500G
2-Amino-4,6-dichloropyrimidine 95%
2-Amino-4,6-dichloropyrimidine
Formule: C₄H₃Cl₂N₃
Smeltpunt: 218…222 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006090
CAS nummer: 56-05-3
EINECS: 200-253-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR