3-Bromo-5-chlorobenzonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 3-Bromo-5-chlorobenzonitile

OR11380-10G OR11380-1G OR11380-5G
APOSOR11380-10GEA 40.9 EUR
APOSOR11380-10G APOSOR11380-1G APOSOR11380-5G
3-Bromo-5-chlorobenzonitrile
3-Bromo-5-chlorobenzonitrile
Formule: C₇H₃BrClN
MW: 216.46 g/mol
Smeltpunt: 70.7…71.1 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06200840
CAS nummer: 304854-55-5
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR