3,5-Dichloropyridine-2-carbonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR11385-25G OR11385-100G OR11385-500G
APOSOR11385-25GEA 39.5 EUR
APOSOR11385-25G APOSOR11385-100G APOSOR11385-500G
3,5-Dichloropyridine-2-carbonitrile
3,5-Dichloropyridine-2-carbonitrile
Formule: C₆H₂Cl₂N₂
Smeltpunt: 98…103 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03788758
CAS nummer: 85331-33-5
UN: 3439
ADR: 6.1,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR