2-Amino-6-chloropyrazine 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR11609-25G OR11609-100G OR11609-5G
APOSOR11609-25GEA 51.2 EUR
APOSOR11609-25G APOSOR11609-100G APOSOR11609-5G
2-Amino-6-chloropyrazine 98%
2-Amino-6-chloropyrazine
Formule: C₄H₄ClN₃
MW: 129.55 g/mol
Smeltpunt: 150…152 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00055024
CAS nummer: 33332-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR