2-Bromoisonicotinonitrile 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR11615-1G OR11615-5G OR11615-25G
APOSOR11615-1GEA 24.9 EUR
APOSOR11615-1G APOSOR11615-5G APOSOR11615-25G
2-Bromoisonicotinonitrile 95%
2-Bromoisonicotinonitrile
Formule: C₆H₃BrN₂
Smeltpunt: 70…74 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07367879
CAS nummer: 10386-27-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR