2-Amino-5-nitropyridine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR1164-100G OR1164-25G OR1164-500G
APOSOR1164-100GEA 29.2 EUR
APOSOR1164-100G APOSOR1164-25G APOSOR1164-500G
2-Amino-5-nitropyridine
2-Amino-5-nitropyridine
Formule: C₅H₅N₃O₂
Smeltpunt: 186…190 °C
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006325
CAS nummer: 4214-76-0
EINECS: 224-145-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR