5-Chlorosalicylaldehyde 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 5-Chloro-2-hydroxybenzaldehyde

OR1167-100G OR1167-25G OR1167-500G
APOSOR1167-100GEA 48.3 EUR
APOSOR1167-100G APOSOR1167-25G APOSOR1167-500G
5-Chlorosalicylaldehyde 97%
5-Chlorosalicylaldehyde
Formule: C₇H₅ClO₂
MW: 156.57 g/mol
Kookpunt: 105
Smeltpunt: 99…103 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003331
CAS nummer: 635-93-8
EINECS: 211-244-4
UN: 3077
ADR: 9+EHS III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR