trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-) 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR1171-100G OR1171-25G OR1171-500G
APOSOR1171-100GEA 27.8 EUR
APOSOR1171-100G APOSOR1171-25G APOSOR1171-500G
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-) 98%
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid
Formule: C₁₀H₁₀O₄
Smeltpunt: 169…173 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004400
CAS nummer: 537-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR