4-Bromo-N,N-diethylbenzamide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR11919-1G OR11919-5G OR11919-25G
APOSOR11919-1GEA 22 EUR
APOSOR11919-1G APOSOR11919-5G APOSOR11919-25G
4-Bromo-N,N-diethylbenzamide
4-Bromo-N,N-diethylbenzamide
Formule: C₁₁H₁₄BrNO
MW: 256.1390 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00463697
CAS nummer: 5892-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR